Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE-CF594 Rat Anti-Mouse MerTK (Mer), BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 570402
Description
MerTK (Mer) Monoclonal antibody specifically detects MerTK (Mer) in Mouse samples. It is validated for Flow cytometry
Specifications
MerTK (Mer) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
BB14-16 | |
PE-CF594 | |
Mer; proto-oncogene c-Mer; Mertk; Eyk; Nyk | |
Recombinant Mouse MerTK Protein | |
25 μg | |
17289 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction