Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE-CF594 Rat Anti-Mouse MerTK (Mer), BD Horizon™
Rat Monoclonal Antibody
$130.60 - $317.00
Specifications
| Antigen | MerTK (Mer) |
|---|---|
| Clone | BB14-16 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
MerTK (Mer) Monoclonal antibody specifically detects MerTK (Mer) in Mouse samples. It is validated for Flow cytometrySpecifications
| MerTK (Mer) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| 17289 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| BB14-16 | |
| Flow Cytometry | |
| PE-CF594 | |
| Rat | |
| Mouse | |
| Mer; proto-oncogene c-Mer; Mertk; Eyk; Nyk | |
| Recombinant Mouse MerTK Protein | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title