Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Mouse Anti-Human Neuropilin-1 (CD304), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570223
Description
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry
Specifications
Neuropilin-1 (CD304) | |
Monoclonal | |
PE | |
BDCA-4; Blood dendritic cell antigen 4; NRP; NRP1; Neuropilin; VEGF165R | |
Human NRP1 b1b2 Ectodomain | |
100 Tests | |
8829 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
mAb3 (also known as MAB3; mAb#3) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction