Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Mouse Anti-Human Neuropilin-1 (CD304), BD Pharmingen™
Mouse Monoclonal Antibody
$144.00 - $332.00
Specifications
Antigen | Neuropilin-1 (CD304) |
---|---|
Clone | mAb3 (also known as MAB3; mAb#3) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometrySpecifications
Neuropilin-1 (CD304) | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
BDCA-4; Blood dendritic cell antigen 4; NRP; NRP1; Neuropilin; VEGF165R | |
Human NRP1 b1b2 Ectodomain | |
Affinity Chromatography |
mAb3 (also known as MAB3; mAb#3) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
8829 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title