Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Mouse Anti-Mouse TCR Vγ6, BD Pharmingen™
Mouse Monoclonal Antibody
$164.00 - $372.00
Specifications
| Antigen | TCR Vγ6 |
|---|---|
| Clone | 1C10-1F7 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
TCR Vγ6 Monoclonal antibody specifically detects TCR Vγ6 in Mouse samples. It is validated for Flow cytometrySpecifications
| TCR Vγ6 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 21640 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 1C10-1F7 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Mouse | |
| TCR Vg6; TCR Vgamma6; TCR Vγ6; TRGV6; TcR Vγ4; Tcrg-V6; Trgv6 | |
| Mouse TCR Vγ6 Peptide | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title