Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Rat Anti-β-galactosidase, BD Pharmingen™
Rat Monoclonal Antibody
$130.00 - $322.00
Specifications
Antigen | β-galactosidase |
---|---|
Clone | 13R4.rMAb (also known as 13R4) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
β-galactosidase Monoclonal antibody specifically detects β-galactosidase in Bacteria samples. It is validated for Intracellular staining (flow cytometry)Specifications
β-galactosidase | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
945006 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
13R4.rMAb (also known as 13R4) | |
Flow Cytometry | |
PE | |
Rat | |
Bacteria | |
beta-gal; beta-galactosidase; lacZ; lactase; β-gal; β-galactosidase | |
E. coli β-galactosidase | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title