Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PE Rat Anti-Human HER4 (ErbB4), BD Pharmingen™

Catalog No. p-200020621 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570218 0.1 mg
BDB570219 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570218 Supplier BD Biosciences Supplier No. 570218
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HER4 (ErbB4) Monoclonal antibody specifically detects HER4 (ErbB4) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HER4 (ErbB4)
Applications Flow Cytometry
Classification Monoclonal
Clone P6-1
Concentration 0.2 mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ERBB4; ErbB4; HER-4; c-ErbB-4; erb-b2 receptor tyrosine kinase 4; erbB-4
Host Species Rat
Immunogen Human HER4-transfected Rat RH7777 Cells
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 2066
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.