Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEAR1 Antibody (492621), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB369881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369881 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369881 Supplier Novus Biologicals Supplier No. FAB4527L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PEAR1 Monoclonal specifically detects PEAR1 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen PEAR1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 492621
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias JEDI, platelet endothelial aggregation receptor 1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human PEAR1, Leu21-Ser754, Accession # Q5VY43
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375033
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.