Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECR Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB097938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097938 Supplier Novus Biologicals Supplier No. NBP297904AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PECR Polyclonal specifically detects PECR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PECR
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias DCR-RP, EC 1.3.1.38, HPDHASE, HSA250303, peroxisomal trans-2-enoyl-CoA reductase, putative short chain alcohol dehydrogenase, pVI-ARL, PVIARL2,4-dienoyl-CoA reductase-related protein, SDR29C1, short chain dehydrogenase/reductase family 29C, member 1, TERPDCRRP
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PECR fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55825
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.