Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEDFR/PNPLA2/ATGL Antibody (1H6), Novus Biologicals™
SDP

Catalog No. NB298229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298229 100 μL
NB298228 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298229 Supplier Novus Biologicals Supplier No. NBP326374100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PEDFR/PNPLA2/ATGL Monoclonal antibody specifically detects PEDFR/PNPLA2/ATGL in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen PEDFR/PNPLA2/ATGL
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 1H6
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Adipose triglyceride lipase, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF-R, Pigment epithelium-derived factor, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2.2, TTS-2.2, TTS2DKFZp667M109
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PEDFR/PNPLA2/ATGL [UniProt Q96AD5]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57104
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.