Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trajan Scientific and Medical PEEK Tubing, 1/8 in. O.D., 10 ft

Click to view available options
Diameter (English) Inner:
0.062 in.
0.080 in.
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
Specifications
Color | Natural |
pH Range | 2 to 14 |
Diameter (English) Inner | 0.062 in. |
Diameter (English) Outer | 1/8 in. |
Overall Length (English) | 10 ft. |
Material | PEEK |
Maximum Pressure | 4000 psi (276 bar) |
Max. Operating Temperature (Metric) | 100°C |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction