Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEG, Mouse anti-All, Clone: 6.3, Millipore Sigma™

Catalog No. MABS1966100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1966100 100 μg
MABS196625 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS1966100 Supplier MilliporeSigma Supplier No. MABS1966100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PEG, clone 6.3, Cat. No. MABS1966, is a mouse monoclonal antibody that detects PEG-conjugated proteins and it hs been tested for use in ELISA.

Polyethylene glycols (PEG) are non-toxic, uncharged, hydrophilic long chain polymers with a big exclusion volume in aqueous solution. The covalent attachment of PEG is commonly employed to modify proteins and drugs that can increase their bioavailability and reduce immunogenicity and proteolytic degradation. Clone 6.3 displays high affinity and binds the PEG backbone and can be employed as a detection antibody for the quantification of PEG. It binds to a smaller epitope of the PEG backbone when compared to other antibodies. Since it binds to a smaller PEG epitope (∽750 Da), it is not compatible with detergents that contain an ethylene oxide backbone, such as Tween 20. Instead 0.05% CHAPS is a more suitable detergent when using this antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEG
Applications ELISA
Classification Monoclonal
Clone 6.3
Conjugate Unconjugated
Dilution ELISA Analysis: A representative lot detected PEG in ELISA applications (Tung, H.Y., et. al. (2015). Mol Cancer Ther. 14(6):1317-26).
Formulation Purified mouse monoclonal antibody IgG1 in PBS with 0.04% sodium azide.
Gene Alias Polyethylene Glycol
Host Species Mouse
Immunogen PEG-conjugated to E.coli beta-glucuronidase.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 6.3 specifically detects proteins conjugated with polyethylene glycol (PEG).
Target Species All species
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.