Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEG, Mouse anti-All, Clone: 6.3, Millipore Sigma™
Mouse Monoclonal Antibody
$209.00 - $525.00
Specifications
| Antigen | PEG |
|---|---|
| Clone | 6.3 |
| Dilution | ELISA Analysis: A representative lot detected PEG in ELISA applications (Tung, H.Y., et. al. (2015). Mol Cancer Ther. 14(6):1317-26). |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABS196625
|
MilliporeSigma
MABS196625UG |
25 μg |
Each for $209.00
|
|
|||||
|
MABS1966100
|
MilliporeSigma
MABS1966100UG |
100 μg |
Each for $525.00
|
|
|||||
Description
Anti-PEG, clone 6.3, Cat. No. MABS1966, is a mouse monoclonal antibody that detects PEG-conjugated proteins and it hs been tested for use in ELISA.
Polyethylene glycols (PEG) are non-toxic, uncharged, hydrophilic long chain polymers with a big exclusion volume in aqueous solution. The covalent attachment of PEG is commonly employed to modify proteins and drugs that can increase their bioavailability and reduce immunogenicity and proteolytic degradation. Clone 6.3 displays high affinity and binds the PEG backbone and can be employed as a detection antibody for the quantification of PEG. It binds to a smaller epitope of the PEG backbone when compared to other antibodies. Since it binds to a smaller PEG epitope (∽750 Da), it is not compatible with detergents that contain an ethylene oxide backbone, such as Tween 20. Instead 0.05% CHAPS is a more suitable detergent when using this antibody.Specifications
| PEG | |
| ELISA Analysis: A representative lot detected PEG in ELISA applications (Tung, H.Y., et. al. (2015). Mol Cancer Ther. 14(6):1317-26). | |
| Monoclonal | |
| Purified | |
| RUO | |
| All species | |
| Polyethylene Glycol | |
| IgG1 κ | |
| Protein A purified | |
| Clone 6.3 specifically detects proteins conjugated with polyethylene glycol (PEG). |
| 6.3 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Signaling | |
| Purified mouse monoclonal antibody IgG1 in PBS with 0.04% sodium azide. | |
| PEG-conjugated to E.coli beta-glucuronidase. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title