Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEG, Mouse anti-All, Clone: AGP4, Millipore Sigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1964100UG
Description
Anti-PEG, clone AGP4, Cat. No. MABS1964, is a mouse monoclonal antibody that detects Polyethylene glycol (PEG) and has been tested for use in ELISA, Western Blotting, and Immunohistochemistry.
Polyethylene glycols (PEG) are non-toxic, uncharged, hydrophilic long chain polymers with a big exclusion volume in aqueous solution. The covalent attachment of PEG is commonly employed to modify proteins and drugs that can increase their bioavailability and reduce immunogenicity and proteolytic degradation. PEG is considered as a non-immunogenic moiety and has been widely tested for its ability to protect proteins from the host s immune system. This antibody (Clone AGP4) binds to the PEG backbone and is suitable for capture of PEGylated compounds in ELISA as well as the detection of PEGylated molecules. It is suitable for sensitive ELISA applications where AGP4 can be used as a capture antibody. Note: Tween-20 should be used in wash buffers for this antibody combination.
Specifications
PEG | |
Monoclonal | |
Unconjugated | |
Purified mouse monoclonal antibody IgM in PBS with 0.04% sodium azide. | |
Mouse | |
Purified by ion-exchange chromatography | |
RUO | |
Primary | |
All species | |
Purified |
ELISA, Immunohistochemistry, Western Blot | |
AGP4 | |
Immunohistochemistry Analysis: A representative lot detected PEG in Immunohistochemistry applications (Wang, H., et. al. (2012). J Virol. 86(11):6286-302).Western Blotting Analysis: A representative lot detected PEG in Western Blotting applications (Ehrlich, G.K., et. al. (2009). J Mol Recognit. 22(2):99-103).ELISA Analysis: A representative lot detected PEG in ELISA applications (Hao WR et al (2018). Sci. Rep. 8(1); 4256; Chang, C.J., et. al. (2017). Nat Commun. 8(1); 522; Henry, C.E., et. al. (2016). Acta Biomater. 43:61-70). | |
Polyethylene Glycol | |
Polyethylene glycol-conjugated to E.coli beta-glucuronidase. | |
100 μg | |
Signaling | |
Clone AGP4 specifically detects proteins conjugated with low to high molecular weight methoxy and amino polyethylene glycols (PEGs). | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction