Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEG10 Antibody (981902), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB295286 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295286 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295286 Supplier Novus Biologicals Supplier No. FAB99751S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

PEG10 Monoclonal antibody specifically detects PEG10 in Human samples. It is validated for ELISA, Flow Cytometry, CyTOF

Specifications

Antigen PEG10
Applications ELISA, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 981902
Conjugate Alexa Fluor 750
Dilution ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias embryonal carcinoma differentiation regulated, Embryonal carcinoma differentiation-regulated protein, HB-1, KIAA1051EDR, Mammalian retrotransposon-derived protein 2, Mar2, Mart2, MEF3 like 1, MEF3L, MEF3L1, Myelin expression factor 3-like protein 1, paternally expressed 10, Paternally expressed gene 10 protein, retrotransposon gag domain containing 3, Retrotransposon gag domain-containing protein 3, Retrotransposon-derived gag-like polyprotein, retrotransposon-derived protein PEG10, RGAG3MEF3-like protein 1, Ty3/Gypsy-like protein
Host Species Mouse
Immunogen Synthetic peptide containing human PEG10, Accession # Q86TG7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23089
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.