Learn More
Eisco™ Pegmatite Igneous Rock Specimen
SCI_ED
Catalog No. s27590
Click to view available options
Quantity:
1/Ea.
12/Pk.
Pegmatite is an igneous rock that contains two vastly different crystal sizes. This combination of both phaneritic and aphanitic crystals indicates this rock formed at two vastly different cooling rates.
  • Correlates well to Next Generation Science Standards Evidence
  • Pegmatite is a classic rock for teaching igneous petrology
  • Useful in volcanology lessons
Specifications
Product Type Igneous Rock
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.