Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PEI STAR-Go™ transfection reagent

Catalog No. 81-745-ML
Click to view available options
Quantity:
5 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Aqueous solution of polyethylenimine (PEI) transfection reagent

PEI STAR-Go™ is a ready-to-use aqueous formulation of our high performance PEI STAR™ transfection reagent (Cat. No. 7854), provided as a sterile-filtered solution. PEI STAR™ is a chemically-defined, linear polyethylenimine (PEI) transfection reagent for affordable and scalable transient gene expression. PEI is a synthetic polymer with an exceptionally high positive charge density in pH-neutral solutions. Positively charged PEI binds strongly to negatively charged DNA and imparts a net cationic charge, allowing the DNA to enter cells. PEI is a non-viral vector commonly used to transfect HEK293, CHO, and a range of other eukaryotic cell lines. Applications include production of lentivirus and rAAV, recombinant proteins, and antibodies. Supplied as a 1mg/mL aqueous solution

PEI STAR-Go is a trademark of Bio-Techne Corp.

Specifications

Chemical Name or Material Poly[imino(1,2-ethanediyl)] hydrochloride
CAS 49553-93-7
Quantity 5 mL
Target Transfection Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.