Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PELI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7207350 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0710 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0710 Supplier Proteintech Group Inc Supplier No. 160971AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Scaffold protein which probably links Toll-like receptors (TLRs) to basic cellular processes via its interaction with the complex containing IRAK kinases and TRAF6. Can activate the MAP (mitogen activated protein) kinase pathway leading to activation of ELK1. Not required for NF-kappa-B activation.

Specifications

Antigen PELI2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PELI2
Gene Accession No. Q8BST6, Q9HAT8
Gene Alias E3 ubiquitin-protein ligase pellino homolog 2, PELI2, Pellino 2, pellino homolog 2 (Drosophila), Protein pellino homolog 2
Gene Symbols PELI2
Host Species Rabbit
Immunogen PELI2 Fusion Protein Ag8940
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305835, 57161, 93834
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.