Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pellino 1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB192280 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB192280 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB192280 Supplier Novus Biologicals Supplier No. NBP281888F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pellino 1 Polyclonal antibody specifically detects Pellino 1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen Pellino 1
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1
Host Species Rabbit
Immunogen Rabbit polyclonal Pellino 1 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human Pellino 1. The immunogen is located within amino acids 300 - 350 of Pellino 1. Amino Acid Squence: CHNWGNKEERDGKDRE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57162
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.