Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PELP1 Rabbit anti-Human, Mouse, Rat, Clone: 9V9A5, Novus Biologicals™
SDP

Catalog No. NB168563 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168563 100 μg
NB168564 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168563 Supplier Novus Biologicals Supplier No. NBP316736100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PELP1 Monoclonal antibody specifically detects PELP1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PELP1
Applications Western Blot
Classification Monoclonal
Clone 9V9A5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias HMX3, MNARmodulator of nongenomic activity of estrogen receptor, Modulator of non-genomic activity of estrogen receptor, P160, proline and glutamic acid rich nuclear protein, proline, glutamate and leucine rich protein 1, proline, glutamic acid and leucine rich protein 1, proline-, glutamic acid- and leucine-rich protein 1, proline-, glutamic acid-, leucine-rich protein 1, Transcription factor HMX3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1000-1100 of human PELP1 (Q8IZL8).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 27043
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.