Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ X Series Forestry Ear Muffs

PELTOR™ X Series Forestry Ear Muffs are electrically insulated with orange coloring to improve visibility for use in the forestry and other applicable industries.

Specifications

Color Orange
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Forestry, Landscaping, Utilities, Mining, Oil and Gas, Electrical Contractors
High Visibility Yes
View More Specs

Products 2
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
19-153-672
Accessories View Documents
3M™
7100097447
X1P5E-OR Hard Hat 21 dB N/A N/A N/A
19-153-671
View Documents
3M™
7100097446
X4A-OR Over-the-head 27 dB N/A N/A N/A
Description

Description

  • Ideal for use in forestry and landscaping industries
  • Electrically insulated
  • Orange color is highly visible
  • High-performance arms to balance durability and flexibility
  • Easy click in/click out
  • Hard hat compatibility (models X1P5E and X4P5E)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Orange
Black
Yes
X Series Forestry Ear Muffs
Foam, Plastic, Metal
Yes
Forestry, Landscaping, Utilities, Mining, Oil and Gas, Electrical Contractors
1 Ea.
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.