Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Pentacarbonyliron, 99.5%
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                25 g
                            
                        
                            
                                1 kg
                            
                        
                    Description
Pentacarbonyliron is used as a catalyst in organic reactions, and as carbonyl iron for high-frequency coils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Melting Point | -20°C | 
| Color | Red | 
| Density | 1.457 g/mL | 
| Boiling Point | 103°C (200°C (decomposition)) | 
| Flash Point | -15°C (5°F) | 
| Odor | Pungent | 
| Refractive Index | 1.5196 | 
| Linear Formula | Fe(CO)5 | 
| Quantity | 25 g | 
| UN Number | UN1994 | 
| Show More | 
Safety and Handling
| Hazard Category | H225-H300+H310+H330-H372 | 
| Hazard Statement | GHS H Statement H225-H300-H310-H330 Highly flammable liquid and vapor. Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled.  | 
                                
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P312-P314-P330-P363-P370+P378q-P501c | 
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: IRON PENTACARBONYL | 
| EINECSNumber | 236-670-8 | 
| RTECSNumber | NO4900000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures; Store under Argon | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction