Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoropropionic acid, 97%

Catalog No. ac416920100
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle

Catalog No. AC416920100

Supplier: Thermo Scientific Chemicals 416920100

Only null left
Add to Cart

CAS: 422-64-0 | C3HF5O2 | 164.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-64-0
Molecular Formula C3HF5O2
Molecular Weight (g/mol) 164.03
MDL Number MFCD00004170
InChI Key LRMSQVBRUNSOJL-UHFFFAOYSA-N
Synonym pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid
PubChem CID 62356
SMILES OC(=O)C(F)(F)C(F)(F)F

Specifications

Color Colorless to Brown
Density 1.5760g/mL
Boiling Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Refractive Index 1.2830 to 1.286
Linear Formula CF3CF2CO2H
Quantity 10 g
Specific Gravity 1.576
IUPAC Name 2,2,3,3,3-pentafluoropropanoic acid
Formula Weight 164.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Pentafluoropropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-021-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.