Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane (Spectranalyzed™), Fisher Chemical

Catalog No. P3931 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P393-1 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P393-1

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109-66-0 , 287-92-3 , 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Density 0.626g/cm³
Quantity 1 L
Flash Point -49°C(120.2°F)
Identification Pass Test
Infrared Spectrum Conforms to reference
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 573mbar at 20°C
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa/s at 20°C
UN Number 1265
Boiling Point 35°C
Formula Weight 72.15
Grade Spectranalyzed
Chemical Name or Material Pentane
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.