Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pentostatin
GREENER_CHOICESDP

Catalog No. 203310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-331-0 10 mg
20-335-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-331-0 Supplier Tocris Bioscience™ Supplier No. 2033/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Adenosine deaminase inhibitor

Irreversible inhibitor of adenosine deaminase (Ki = 2.5 pM). Anticancer agent.

Chemical Identifiers

CAS 53910-25-1
Molecular Formula C11H16N4O4

Specifications

Quantity 10 mg
Formula Weight 268.27
Purity >99%
Molecular Weight (g/mol) 268.27
Chemical Name or Material Pentostatin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.