Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentraxin 3/TSG-14 Antibody (MM0512-6B7) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP211849 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211849 0.1 mg
NB018922 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP211849 Supplier Novus Biologicals Supplier No. NBP211849

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Pentraxin 3/TSG-14 Monoclonal specifically detects Pentraxin 3/TSG-14 in Human samples. It is validated for Western Blot.

Specifications

Antigen Pentraxin 3/TSG-14
Applications Western Blot
Classification Monoclonal
Clone MM0512-6B7
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. P26022
Gene Alias alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14
Gene Symbols PTX3
Host Species Mouse
Immunogen Human recombinant Pentraxin 3/TSG-14
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5806
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.