Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEPP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP237696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237696 Supplier Novus Biologicals Supplier No. NBP237696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PEPP2 Polyclonal specifically detects PEPP2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PEPP2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_061885.3
Gene Alias FLJ10667, FLJ26734, KIAA1686FLJ31492, PEPP-2, PEPP2phosphoinositol 3-phosphate-binding protein-2, PH domain-containing family A member 5, Phosphoinositol 3-phosphate-binding protein 2, pleckstrin homology domain containing, family A member 5, pleckstrin homology domain-containing family A member 5
Gene Symbols PLEKHA5
Host Species Rabbit
Immunogen This antibody maps to a region between residue 876 to 925 of human Pleckstrin Homology Domain Containing, Family A Member 6 using the numbering given in entry NP_061885.3 (GeneID 54477).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54477
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.