Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pepsin, MP Biomedicals™

Catalog No. MP021953672 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021953672 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021953672 Supplier MP Biomedicals™ Supplier No. 02195367.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to digest proteins and in the preparation of Fab fragments from antibodies. Pepsin, MP Biomedicals is an acid protease that contains a proteolytic enzyme with a in.lcathepsinin.l component that has milk curdling activity. It has a broad range of substrate

Pepsin is the major proteolytic enzyme produced in the stomach and is used to digest proteins. It is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice.

Pepsin digests proteins through the cleavage of interior peptide linkages. It hydrolyzes peptide bonds, not amide or ester linkages.

  • Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond
  • Sulfur-containing amino acids increase susceptibility to hydrolysis when they are close to the peptide bond
  • Pepsin preferentially cleaves at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid residues
  • Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowy Tan
pH Optimum pH 1 to 3.5, unstable above 6.0
For Use With (Application) Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice.
Content And Storage -20°C, Desiccate
Source Porcine Stomach Mucosa
Quantity 250 mg
Molecular Weight (g/mol) ∽35,000
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.