Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pepsinogen A4/PGA4 Antibody (10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB383214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383214 Supplier Novus Biologicals Supplier No. NBP312536AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pepsinogen A4/PGA4 Monoclonal antibody specifically detects Pepsinogen A4/PGA4 in Human samples. It is validated for ELISA

Specifications

Antigen Pepsinogen A4/PGA4
Applications ELISA
Classification Monoclonal
Clone 10
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen A4/PGA4 (Accession#: NP_001073276.1; Met1-Ala388).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643847
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.