Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pepsinogen C/PGC/Progastricsin Antibody (90/11), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1062819A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062819A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062819A30 Supplier Novus Biologicals Supplier No. NB10062819AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pepsinogen C/PGC/Progastricsin Monoclonal specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA.

Specifications

Antigen Pepsinogen C/PGC/Progastricsin
Applications ELISA
Classification Monoclonal
Clone 90/11
Conjugate Alexa Fluor 350
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C)
Gene Symbols PGC
Host Species Mouse
Immunogen Human pepsinogen II
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5225.0
Test Specificity NB100-62819 is specific for human pepsinogen II. Less than 1% reactivity has been observed with human pepsinogen I (isozymogens 2-5), porcine pepsinogen, gastrin I, somatostatin, peptide YY and bombesin. Affinity: 1 x 109 l/mol
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.