Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pepsinogen C/PGC/Progastricsin Antibody (90/11), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10062819G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062819G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062819G Supplier Novus Biologicals Supplier No. NB10062819G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pepsinogen C/PGC/Progastricsin Monoclonal specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA.

Specifications

Antigen Pepsinogen C/PGC/Progastricsin
Applications ELISA
Classification Monoclonal
Clone 90/11
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C)
Gene Symbols PGA3
Host Species Mouse
Immunogen Human pepsinogen II
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643834
Test Specificity NB100-62819 is specific for human pepsinogen II. Less than 1% reactivity has been observed with human pepsinogen I (isozymogens 2-5), porcine pepsinogen, gastrin I, somatostatin, peptide YY and bombesin. Affinity: 1 x 109 l/mol
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.