Learn More
Pepsinogen C/PGC/Progastricsin Antibody, Alexa Fluor™ 700, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Pepsinogen C/PGC/Progastricsin |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C) |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Pepsinogen C/PGC/Progastricsin (NP_579818.1; Met1-Val392). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.