Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pepsinogen I Antibody (8003 (99/12)), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10062239C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062239C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062239C Supplier Novus Biologicals Supplier No. NB10062239C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pepsinogen I Monoclonal specifically detects Pepsinogen I in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA).

Specifications

Antigen Pepsinogen I
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA)
Classification Monoclonal
Clone 8003 (99/12)
Conjugate DyLight 650
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias pepsinogen 3, group I (pepsinogen A)
Gene Symbols PGA3
Host Species Mouse
Immunogen Purified human pepsinogen 1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643834
Test Specificity NB100-62239 recognises human Pepsinogen I, a zymogen or proenzyme secreted by Chief cells in the stomach. It is cleaved to form pepsin both in an autocatalytic fashion and by pepsin itself. In humans there are two related forms of pepsin, Pepsinogen I (also known as pepsinogen A), and Pepsinogen II (also known as Pepsinogen B or progastricsin).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.