Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEPT1/SLC15A1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB127153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB127153 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB127153 Supplier Novus Biologicals Supplier No. NBP298544FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PEPT1/SLC15A1 Polyclonal specifically detects PEPT1/SLC15A1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PEPT1/SLC15A1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Caco-2 oligopeptide transporter, HPECT1macrophage oligopeptide transporter PEPT1, HPEPT1, Intestinal H(+)/peptide cotransporter, Oligopeptide transporter, small intestine isoform, PEPT1intestinal H+/peptide cotransporter, Peptide transporter 1, solute carrier family 15 (oligopeptide transporter), member 1, solute carrier family 15 member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PEPT1/SLC15A1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6564
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.