Learn More
Description
Specifications
Specifications
| Antigen | PER2 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation reported in scientific literature (PMID 31390562), Immunohistochemistry-Paraffin 1:200. Use reported in scientific literature (PMID 31822134) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O15055 |
| Gene Alias | Circadian clock protein PERIOD 2, FASPS, hPER2, KIAA0347period 2, period (Drosophila) homolog 2, period circadian protein 2, period circadian protein homolog 2, period homolog 2 (Drosophila) |
| Gene Symbols | PER2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
