Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 0.154 Normal in Acetic Acid, Reagents

Catalog No. 01334300
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-300 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-300 Supplier Reagents Holdings Llc Supplier No. CP1205002.5D1
Only null left
Add to Cart
Add to Cart

Perchloric Acid, 0.154 Normal in Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 64-19-7 , 108-24-7 , 7601-90-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide in Glacial Acetic Acid
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH <2
Color Colorless to Light Yellow
Boiling Point 118°C
Packaging Glass Bottle
Specific Gravity 1.0587
Quantity 2.5 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.154 Normal in Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.