Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perchloric Acid, 60 to 62%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
PVC Coated Glass Bottle
Specifications
Specifications
Packaging | PVC Coated Glass Bottle |
Specific Gravity | 1L = 1.50kg |
Quantity | 2.5 L |
Formula Weight | 100.46 |
Percent Purity | 60 to 62% |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | Perchloric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction