Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 69 to 72%, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003547 Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
PVC Coated Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    For Trace Metal Analysis

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Packaging PVC Coated Glass Bottle
Specific Gravity 1L = 1.50kg
Quantity 2.5 L
Formula Weight 100.46
Percent Purity 69 to 72%
Grade BAKER INSTRA-ANALYZED™ Reagent
Chemical Name or Material Perchloric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.