Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071936 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L

ACS Reagent, 70%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Concentration 70%
Density 1.664g/mL at 25°C
Residue after Ignition ≤0.003%
Vapor Pressure 6.8mmHg (25°C)
Flash Point 113°C (235.4°F)
Quantity 100 mL
Linear Formula HClO4
UN Number 1873
Formula Weight 100.46g/mol
Grade ACS Reagent
Chemical Name or Material Perchloric Acid
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375
Precautionary Statement H271-H290-H302-H314-H373
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.