Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Percoll™, MP Biomedicals™

Catalog No. mp219536980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219536980 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219536980 Supplier MP Biomedicals, Inc Supplier No. 0219536980
Only null left
Add to Cart
Add to Cart

Composed of colloidal silica coated with polyvinylpyrrolidone (PVP). Percoll™, MP Biomedicals is a medium for density gradient centrifugation of cells, viruses and subcellular particles. It is completely non-toxic and ideal for use with biological m

Centrifugation of Percoll™ results in spontaneous formation of a density gradient due to the heterogeneity of particle sizes in the medium. Percoll™ can be used for formation of gradients either by the use of conventional gradient mixers or by high speed centrifugation. In the latter case the sample can be premixed with the medium and then separated on the gradient created in situ. In this way gradient formation and sample separation can be achieved in a single operation.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65455-52-9

Specifications

Density 1.128g/mL (Lit.)
Quantity 100 mL
Viscosity Record cp at 20°C
Physical Form Liquid
Recommended Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.