Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Percoll™ PLUS Centrifugation Media
GSA_VA

Catalog No. p-3753328 Shop All Cytiva Products
Click to view available options
Quantity:
250 mL
1 L

Catalog No. 45-001-754

Supplier: Cytiva 17544502

Only null left
Add to Cart
Edge
Add to Cart

Low endotoxin reagent for density gradient centrifugation of cells, viruses, and subcellular particles. Cytiva™ Percoll™ PLUS Centrifugation Media is non-toxic, shows low viscosity and is ideal for density gradients.

  • Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
  • Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
  • Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
  • Nontoxic, almost chemically inert and does not adhere to membranes
  • Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
  • Silica sol coated with covalently linked silane; 15-30nm dia. bead size
  • Low endotoxin levels (max. 2 EU/mL); well-suited for cell separation in clinical research applications

For in vitro research use only.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 mL
Content And Storage 4–30°C
For Use With (Application) Cell Isolation
Viscosity 15cP
Endotoxin Level 2 EU/mL
Osmolality < 30 mOsm/kg
Color Colorless to Slightly Yellow
pH 8.9 to 9.9
Product Type Centrifugation Media
Density 1.135 g/mL
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.