Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Antibody (1031751), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB305695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305695 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305695 Supplier Novus Biologicals Supplier No. FAB103853MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Perforin
Applications Flow Cytometry
Classification Monoclonal
Clone 1031751
Conjugate mFluor Violet 450 SE
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line CHO-derived human Perforin, Pro22-Trp555, Accession # P14222
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5551
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.