Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Antibody (B-D48), FITC, Novus Biologicals™
SDP

Catalog No. NB388543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388543 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388543 Supplier Novus Biologicals Supplier No. NBP314581F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry

Specifications

Antigen Perforin
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone B-D48
Conjugate FITC
Formulation PBS
Gene Alias Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Host Species Mouse
Immunogen Natural human Perforin
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5551
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.