Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Antibody (BOR21) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018521 25 μg
NBP14310601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018521 Supplier Novus Biologicals Supplier No. NBP14310625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for Western Blot.

Specifications

Antigen Perforin
Applications Western Blot
Classification Monoclonal
Clone BOR21
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Gene Alias Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Gene Symbols PRF1
Host Species Mouse
Immunogen The immunogen for this antibody was Perforin.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5551
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.