Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Antibody (PRF1/2470), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB011831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011831 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011831 Supplier Novus Biologicals Supplier No. NBP279886AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Perforin
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PRF1/2470
Conjugate Alexa Fluor 594
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Gene Symbols PRF1
Host Species Mouse
Immunogen Recombinant human Perforin protein fragment (around aa 413-552) (exact sequence is proprietary) (Uniprot: P14222)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.