Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Antibody (PRF1/7077R), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB155811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB155811 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB155811 Supplier Novus Biologicals Supplier No. NBP314154IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Perforin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PRF1/7077R
Conjugate DyLight 755
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Host Species Rabbit
Immunogen Recombinant human Perforin protein fragment (around aa 413-552) (exact sequence is proprietary) (Uniprot: P14222)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5551
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.