Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Mouse anti-Human, Non-human Primate, Clone: Pf-80/164, Novus Biologicals™
SDP

Catalog No. NB171933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2000 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171933 2000 μg
NB171934 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171933 Supplier Novus Biologicals Supplier No. NBP3182962000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Perforin Monoclonal antibody specifically detects Perforin in Human, Non-human Primate samples. It is validated for ELISA, ELISPOT

Specifications

Antigen Perforin
Applications ELISA
Classification Monoclonal
Clone Pf-80/164
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Gene Alias Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Host Species Mouse
Immunogen Native human Perforin
Purification Method Protein G purified
Quantity 2000 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5551
Target Species Human, Non-human Primate
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.