Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perforin Mouse anti-Human, Non-human Primate, Clone: Pf-80/164, Novus Biologicals™

Mouse Monoclonal Antibody
$320.50 - $1147.00
Specifications
Antigen | Perforin |
---|---|
Clone | Pf-80/164 |
Concentration | 1 mg/ml |
Applications | ELISA |
Classification | Monoclonal |
Description
Perforin Monoclonal antibody specifically detects Perforin in Human, Non-human Primate samples. It is validated for ELISA, ELISPOTSpecifications
Perforin | |
1 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Non-human Primate | |
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
Native human Perforin | |
Primary | |
Store at 4°C. |
Pf-80/164 | |
ELISA | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
0.2 um filtered PBS | |
5551 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title