Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Performance Plastic Chemical Wash Bottles with 1 Color Label
SCI_ED

Catalog No. S27175 Shop All Eisco Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Soap
Sodium Hypochlorite (Bleach)
Water
Quantity:
1/Pk.
2/Pk.
6/Pk.

Performance Plastic Wash Bottles are made of low-density polyethylene (LDPE) for superior quality, strength, and flexibility. Chemical labels feature the chemical information as well as hazard, safety, and first aid precautionary statements.

  • Chemical specific wash bottles with color coded caps and labels
  • Performance plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won't fade, crack, or peel
  • Sodium Hypochlorite (bleach) wash bottles are opaque (white) to protect contents from UV light rays
  • Other bottles are clear/transparent
  • Bottle labels comply with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS)
  • Bottle Labels also comply with OSHA's HazCom 2012 requirements and Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity (Metric) 1000 mL
Capacity (English) 33.81 oz.
Color Clear
Description Wide mouth
Includes Built in nozzle
Material LDPE
Length (Metric) 6.98 cm
No. per Pack 1
Quantity 1/Pk.
Sterility Non-sterile
Weight (Metric) 77.11 g
Weight (English) 0.17 lb.
Width (Metric) 12.7 cm
Certifications Hazard Communication Standard 29 CFR 1910.1200
Closure Color Clear
Closure Included Yes
Autoclavable Not Autoclavable
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 23.49 cm
Product Type Wash Bottle
For Use With (Equipment) Distilled Water
Width (English) 5 in.
Length (English) 2.75 in.
Height (English) 9.25 in.
Chemical Label Distilled Water
Closure Material Plastic
Closure Size 6.5 cm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.