Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Perhexiline maleate, >99%
GREENER_CHOICESDP

Catalog No. 516610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-661-0 10 mg
51-665-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-661-0 Supplier Tocris Bioscience™ Supplier No. 5166/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carnitine palmitoyltransferase 1 and 2 (CPT1/2) inhibitor

Carnitine palmitoyltransferase 1 and 2 (CPT1/2) inhibitor. Inhibits palmitate oxidation in rat cardiomyocytes and increases cardiac efficiency in isolated rat hearts. Antianginal.

Chemical Identifiers

CAS 6724-53-4
Molecular Formula C23H39NO4

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight 393.56
Purity >99%
Molecular Weight (g/mol) 393.568
Chemical Name or Material 2-(2,2-Dicyclohexylethyl)piperidine maleate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.